Microcentrifuge tubes, 1,7 ml, SuperClear
211-0033EA 54.9 EUR
VWRI211-0033
Microcentrifuge tubes, 1,7 ml, SuperClear
Ultra-clear PP, graduated, autoclavable and freezable down to –80 °C, with flat cap.
  • Can withstand centrifugation to 15000×g (sterile) or 20000×g (non sterile)
  • Fit all standard, high capacity rotors
  • Flat cap can be easily labelled and pierced, including a special 90° hinge, making it easier to close while pipetting one handed
  • Every lot is tested and guaranteed DNase- and RNase-free
Balení: 50 microtubes/sterile bag (sterile version).