You Searched For: Tongs
60  results were found
SearchResultCount:"60"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: RSG
Description: These beaker/cup tongs are made from electrolytically polished stainless steel.
Catalog Number: (ENTEC40-0315)
Supplier: ENTEGRIS INC
Description: ETFE, white.
UOM: 1 * 4 items
Catalog Number: (BOCH2281)
Supplier: BOCHEM
Description: This test tube holder is made of nickel plated steel. For tubes with a diameter from 10 to 25 mm.
UOM: 1 * 1 items
Catalog Number: (RSGA043.290)
Supplier: RSG
Description: Stainless steel 18/10. All-purpose beaker tongs with rubber protected tips. With second diameter for holding small flasks and beakers.
UOM: 1 * 1 items
Supplier: RSG
Description: Stainless steel, electro-polished.
Catalog Number: (BOCH2283)
Supplier: BOCHEM
Description: This test tube holder is made from stainless steel (18/10) and features a wooden handle. For tubes with a diameter from 10 to 25 mm.
UOM: 1 * 1 items
Catalog Number: (231-4145)
Supplier: RSG
Description: Stainless steel ¹⁸/₁₀, anti-magnetic. For easy, safe handling of beakers and similar containers. The jaws are wrapped in ceramic fibres to provide the...
UOM: 1 * 1 items
Catalog Number: (231-0635)
Supplier: RSG
Description: Stainless steel 18/10. Beaker tongs with rubber protected tips. Strong version.
UOM: 1 * 1 items
Supplier: NABERTHERM GMBH
Description: These tongs are used for the easy loading and unloading of furnaces.
Catalog Number: (BOCH2282)
Supplier: BOCHEM
Description: This test tube holder is made from stainless steel (18/10). For tubes with a diameter from 10 to 25 mm.
UOM: 1 * 1 items
Catalog Number: (USBE4110)
Supplier: USBECK Laborgeräte
Description: Stainless steel beaker tongs with vinyl coated tips.
UOM: 1 * 1 items
Catalog Number: (RSGA043.230)
Supplier: RSG
Description: These beaker tongs are made from electrolytically polished stainless steel.
UOM: 1 * 1 items
Catalog Number: (RSGA80.043.330)
Supplier: RSG
Description: Beaker/cup tongs are made from stainless steel 1.4301.
UOM: 1 * 1 items
Supplier: BOCHEM
Description: Stainless steel ¹⁸/₁₀, anti-magnetic and electropolished. For easy, safe handling of crucibles made from a wide variety of materials.
Catalog Number: (BOCH2110)
Supplier: BOCHEM
Description: Nickel.
UOM: 1 * 1 items
Supplier: BOCHEM
Description: Crucible tongs with ridges, made from stainless steel.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 60
no targeter for Bottom