You Searched For: Staining Tiles
9  results were found
SearchResultCount:"9"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: GLASWARENFABRIK KARL HECHT
Description: Glass spot plates with 10 or 12 numbered cavities. For flocculation tests.
Supplier: MARIENFELD
Description: Staining tile, Float glass, Transparent, W×D×H: 130×100×6 mm, No. of wells: 12, Well size: Ø×H: 20/22×2 mm
Catalog Number: (631-9143)
Supplier: KARTELL
Description: This PE staining tile has three round depressions of Ã˜ 21 mm.
UOM: 1 * 10 items
Catalog Number: (631-9132)
Supplier: KARTELL
Description: PS. This thin plate is suitable for staining applications.
UOM: 1 * 1 items
Catalog Number: (631-0833)
Supplier: GLASWARENFABRIK KARL HECHT
Description: Glass plates with plain or frosted surface. Individually wrapped.
UOM: 1 * 1 items
Supplier: MORGAN ADVANCED MATERIALS HALD
Description: Glazed porcelain. These large tiles with cavities/wells are suitable for staining applications.
Catalog Number: (BRND473500)
Supplier: Brand
Description: Staining tile, Soda-lime glass, Clear, W×D×H: 130×100×6 mm, No. of wells: 12, Well size: Ø: 20 - 22 mm
UOM: 1 * 1 items
Catalog Number: (MARI1800002)
Supplier: MARIENFELD
Description: Staining tile, Glass, W×D×H: 89×57×4,5 mm, No. of wells: 12, Well size: Ø×H: 15×1,5 mm
UOM: 1 * 12 items
Catalog Number: (VWRU89038-192)
Supplier: VWR Collection
Description: Porcelain staining plate, glased (except for bottom surface).
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 9 of 9
no targeter for Bottom