You Searched For: Stirring Rods
24  results were found
SearchResultCount:"24"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Solid PTFE.
Supplier: VWR Collection
Description: PTFE with steel core.
Supplier: Schott AG
Description: Very long glass rods made from Borosilicate 3.3 glass.
Supplier: Thermo Scientific
Description: The fused quartz rods are manufactured by fusing naturally occurring crystalline silica.
Catalog Number: (231-0074)
Supplier: Thermo Fisher Scientific
Description: Stirring rod with flattened ends forming flat paddle 16×16 mm at one end and triangular paddle at the other end.
UOM: 1 * 12 items
Catalog Number: (441-0217)
Supplier: Brand
Description: Disposable stirring spatulas made from PS.
UOM: 1 * 500 items
Catalog Number: (441-0099)
Supplier: KARTELL
Description: For various laboratory uses.
UOM: 1 * 100 items
Catalog Number: (HACH177001)
Supplier: Hach
Description: Stirring rods, Stirring rod, Glass, Length: 150 mm, Ø: 5 mm
UOM: 1 * 3 items
Catalog Number: (300-0517)
Supplier: Sampling Systems
Description: The hand paddle with holes is perfect for stirring and mixing liquids.
UOM: 1 * 1 items
Supplier: GLASWARENFABRIK KARL HECHT
Description: AR-Glas®. With fused ends.
Catalog Number: (442-4160)
Supplier: KARTELL
Description: White, one-piece rod with flattened ends which form two small spatulas.
UOM: 1 * 10 items
Supplier: Corning
Description: These PS stirrer are ideal for use in the dairy, environmental, microbiology and industry sectors. Colour: white.
Supplier: Bel-Art Products, a Part of SP
Description: Assorted shapes for stirring and mixing.
Supplier: DWK Life Sciences
Description: AR-Glas® with fused ends.
Catalog Number: (GERB5430)
Supplier: GERBER FUNKE DR N.
Description: Pestle type stirrer made of glass.
UOM: 1 * 1 items
Supplier: Brand
Description: Stirring rods, Stirring rod, with spatula ends, PTFE, Length: 200 mm, Ø: 8 mm
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 24
no targeter for Bottom