You Searched For: Magnetic Stirring Bars
112  results were found
SearchResultCount:"112"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: PTFE covered, strong Alnico V magnetic core. Smooth surface. For containers with slightly uneven bases.
Supplier: VWR Collection
Description: PTFE coated, strong Alnico V magnetic core. Round, smooth surface. For use in the smallest containers.
Supplier: VWR Collection
Description: PTFE covered, strong Alnico V magnetic core. Rounded, smooth surface.
Supplier: VWR Collection
Description: PTFE covered, strong Alnico V magnetic core. Smooth surface. For containers with slightly uneven bases.
Supplier: VWR Collection
Description: PTFE covered, strong Alnico V magnetic core. Smooth surface. For use in containers with bases that are slightly uneven or curved.
Supplier: VWR Collection
Description: PTFE covered, rare earth magnetic core. The magnetic core is made from a combination of samarium and cobalt. The stirrers will continue to func...
Supplier: VWR Collection
Description: PTFE covered, strong Alnico V magnetic core. Smooth surface. Particularly effective for dissolving solids and mixing sediments because of the s...
Catalog Number: (442-0502)
Supplier: VWR Collection
Description: PTFE covered, strong Alnico V magnetic core. Designed for agitating in 10 mm standard cuvettes.
UOM: 1 * 10 items
Supplier: VWR Collection
Description: PTFE covered, strong Alnico V magnetic core. Smooth surface with rounded edges that intensify the stirring effect, even at low speeds.
Supplier: VWR Collection
Description: PTFE covered, strong Alnico V magnetic core. Smooth surface. Particularly suitable for small stirring vessels, extremely effective stirring act...
Supplier: VWR Collection
Description: PTFE covered, strong Alnico V magnetic core. Smooth surface. Very good centring, small footprint and high turbulence even at low speeds. Disc d...
Supplier: VWR Collection
Description: PTFE covered, Alnico V magnetic core. Smooth surface. For stirring in test tubes, centrifuge tubes and micro vials. The wings can also be trimm...
Supplier: VWR Collection
Description: Set Octahedral 1, PTFE covered, 12 octagonal magnetic stirring rods with centre ring, in practical, transparent box. Set includes two st...
Supplier: VWR Collection
Description: PTFE covered, Alnico V magnetic core. Smooth surface. Designed for use in round bottomed flasks.
Supplier: VWR Collection
Description: PTFE covered, strong Alnico V magnetic core. Smooth surface. Stable rotating position for optimal stirring effect.
Supplier: VWR Collection
Description: PTFE covered, strong Alnico V magnetic core. Smooth surface with rounded edges which enhance agitation effect even at low speeds. Inexpensive, ...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 112
no targeter for Bottom