Safety spectacles, uvex skyper sx2/uvex skybrite sx2
Supplier: Uvex
uvex skybrite sx2
111-1591EA 54.7 EUR
111-1591 111-8006 111-1576 111-1235 111-1236 111-1590
Safety spectacles, uvex skyper sx2/uvex skybrite sx2
Safety Glasses
These coloured, translucent framed, safety spectacles feature a modern design providing an excellent, panoramic field of vision. The arms can be adjusted for length (5 positions) and also for lens inclination. The lens can be easily replaced.
  • Lateral protection
  • Padding at the ends of the arms for increased comfort
  • Special coating makes cleaning easy
Weight: 52 g
Certifikace: EN 166, EN 170; 1F.