Micro powder spatula set
Supplier: RSG Solingen
231-0196EA 28 EUR
231-0196
Micro powder spatula set
Spatulas
Stainless steel 18/10 blade with black plastic handle.
  • Four-piece set, each with a 3, 4, 5 and 6 mm wide powder spatula
  • Blade has excellent chemical and thermal resistance
  • Blades are semicircular and angled