You Searched For: Signs
104  results were found
SearchResultCount:"104"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Brady
Description: Sign, Fire equipment sign, white on red, PE, self-adhesive, 203×203 mm
Catalog Number: (BRDY104400)
Supplier: Brady
Description: ToughStripe™ products are used for marking aisles, passageways and storage locations, and pointing out important safety equipment and egress paths.
UOM: 1 * 1 Roll
Supplier: Brady
Description: A comprehensive range of Emergency Escape and First Aid signs which help comply with current legislation.
Supplier: Brady
Description: Safety signs for the purposes of accident prevention, health hazard information.
Supplier: Brady
Description: Attachable signs warn against particular hazards and provide good safety lighting for dark areas. Barrier chains can be attached to the cones.
Supplier: Brady
Description: All signs comply with current European legislation.
Supplier: Brady
Description: All signs comply with current European legislation.
Supplier: Brady
Description: All signs comply with current European legislation.
Supplier: Brady
Description: Brady B-7541 is a surface printed white polyester film with a permanent, acrylic based, pressure sensitive adhesive and over-laminated with a clear po...
New Product
Supplier: Brady
Description: Safety communication is vital in workplace accident prevention. Instructing and reminding employees and visitors that a laboratory coat must be worn i...
Supplier: Brady
Description: Safety communication is vital in workplace accident prevention. Instructing and reminding employees and visitors that a laboratory coat must be worn i...
Supplier: Brady
Description: Warning signs must have a black pictogram on a yellow triangle with a black border. Hazard signs must be displayed to warn people to be careful or to ...
Supplier: Brady
Description: A range of signs to clearly mark the location of fire extinguishers and other fire fighting equipment as well as fire alarm points.
Supplier: Brady
Description: Standard mandatory signs have always a blue circular backround which contains a white pictogram. These signs display a message or action that must be ...
Supplier: Brady
Description: These PP signs are designed for identification of hazardous substances.
Supplier: Brady
Description: Signs are made of surface printed anti-skid laminated white polyester with a permanent acrylic pressure-sensitive rubber adhesive. They are overlamina...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 104
no targeter for Bottom