You Searched For: Retort Rods
21  results were found
SearchResultCount:"21"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Stainless steel, hollow.
Catalog Number: (444-0919)
Supplier: VWR Collection
Description: Heavy-duty, top-of-the-line rods provide maximum strength and durability. Constructed of 303 stainless steel.
UOM: 1 * 1 items
Catalog Number: (241-0066)
Supplier: VWR Collection
Description: Duralumin, solid.
UOM: 1 * 5 items
Supplier: Bohlender
Description: Virginal rods made of white PTFE.
Supplier: USBECK Laborgeräte
Description: Extension rods made from stainless steel.
Supplier: USBECK Laborgeräte
Description: Stainless steel.
Supplier: USBECK Laborgeräte
Description: Steel, zinc plated.
Supplier: BOCHEM
Description: Stainless steel 18/10.
Supplier: NICKEL ELECTRO
Description: Aluminium, mild steel or stainless steel.
Supplier: BOCHEM
Description: Stainless steel 18/10.
Supplier: BOCHEM
Description: Anodised aluminium.
Supplier: BOCHEM
Description: Tempered cast iron. For 26,9 mm tubes (¾").
Supplier: BOCHEM
Description: Tubes are made of 18/10 stainless steel with grinded surface.
Supplier: BOCHEM
Description: Galvanised steel.
Supplier: BOCHEM
Description: Galvanised steel.
Supplier: BOCHEM
Description: Aluminium, bare.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 21
no targeter for Bottom