You Searched For: Retort Rings
14  results were found
SearchResultCount:"14"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: USBECK Laborgeräte
Description: Wire (Ø 4 mm), powder-coated.
Supplier: USBECK Laborgeräte
Description: Galvanised steel (8 mm Ø).
Supplier: USBECK Laborgeräte
Description: Steel, zinc plated.
Supplier: BOCHEM
Description: These support clips are coated with ceramic inside.
Supplier: BOCHEM
Description: Steel, zinc plated.
Supplier: BOCHEM
Description: Stainless steel 18/10.
Supplier: BOCHEM
Description: Retort rings made from stainless steel 18/10.
Supplier: BOCHEM
Description: Galvanised steel.
Supplier: BOCHEM
Description: Stainless steel 18/10.
Catalog Number: (BOCH5505)
Supplier: BOCHEM
Description: Crucible retort ring made of stainless steel. Ideal for clamping of crucibles for heating substances over a gas flame.
UOM: 1 * 1 items
Supplier: USBECK Laborgeräte
Description: Galvanised steel (8 mm Ø).
Catalog Number: (452-0014)
Supplier: NICKEL ELECTRO
Description: Ring set, Aluminium, Rings with center cover for use with 452-0012
UOM: 1 * 1 items
Supplier: JUCHHEIM Laborgerate GmbH
Description: Steel, epoxy coated.
Supplier: MOHR & CO
Description: Ring, Support ring for 250 ml heating mantles
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 14 of 14
no targeter for Bottom