You Searched For: Specific Gravity Bottles
19  results were found
SearchResultCount:"19"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Brand
Description: Borosilicate glass 3.3, DIN ISO 3507, Gay-Lussac type.
Supplier: Brand
Description: Borosilicate glass 3.3, Gay-Lussac type
Supplier: Brand
Description: Borosilicate glass 3,3, DIN ISO 3507, Gay-Lussac type.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: Pycnometer head only
Supplier: Industrial Physics
Description: Specific gravity cups for determining the specific gravity or density (or weight per gallon wpg) of coatings, pastes or similar liquids.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: Pycnometer, Gay-Lussac, unadjusted, Volume (nom.): 25 ml, Borosilicate glass 3.3
Supplier: Lenz Laborglas GmbH & CO.KG
Description: Borosilicate glass 3.3. Duran®, DIN 12809, ISO 3507, Gay-Lussac type.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: Borosilicate glass 3.3. DURAN®, Gay-Lussac type.
Supplier: GLASWARENFABRIK KARL HECHT
Description: Pycnometer, made of borosilicate glass 3.3, Gay-Lussac type.
Supplier: GLASWARENFABRIK KARL HECHT
Description: Pycnometer, made of borosilicate glass 3.3, Gay-Lussac type.
Catalog Number: (KAVA632431722019)
Supplier: BOHEMIA CRISTAL HANDELSGES.MBH
Description: SIMAX®, borosilicate glass.
UOM: 1 * 10 items
Catalog Number: (WITG8.126.139)
Supplier: WITEG LABORTECHNIK GMBH
Description: Pycnometer, according to Hubbard
UOM: 1 * 1 items
Catalog Number: (SCERSP15115-24)
Supplier: DWK Life Sciences
Description: KIMAX® bottle is used to determine the specific gravity of cement, sand, and other fine materials (ASTM C188).
UOM: 1 * 1 items
Catalog Number: (WITG3.902.002)
Supplier: WITEG LABORTECHNIK GMBH
Description: Made of borosilicate glass.
UOM: 1 * 1 items
Supplier: SciLabware
Description: Borosilicate glass 3.3, Gay-Lussac type.
Supplier: WITEG LABORTECHNIK GMBH
Description: Pycnometer, made of borosilicate glass 3.3, Gay-Lussac type.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 19
no targeter for Bottom