About this item
Finntip™ pipette tips are designed to achieve the highest level of performance in terms of accuracy and repeatability in conjunction with Finnpipette™ pipettes.
- High quality PP, autoclavable
- Unique shape of the collar ensures a leak-free connection to the pipette
- Smooth, hydrophobic surface minimises liquid retention allowing total dispensing of the sample
- Colour coded racks for rapid identification of the tips to be used
- Sterile tips: Certified RNase-, DNase-, DNA- and endotoxin-free
Specifications
- Catalog No:
- 613-4484
- 612-6353
- 613-2608
- 613-2606
- 613-5009
- 613-2605
- 613-2604
- 613-2622
- 613-2603
- 613-7439
- 613-7438
- 613-4485
- 613-5010
- 612-6343
- 613-2599
- 613-2598
- 613-2597
- 613-2615
- 612-6378
- 613-2614
- 613-2595
- 613-2613
- 613-2612
- 612-2508
- 613-7437
- 612-6346
- 613-2601
- 613-2600
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
Specifications
Finntip™ 10, colour code: Pink
Product Information
- Catalog #Supplier #SeqVolumeSterileLengthColour codePackedPkAvailabilityPrice
- 613-25999400300020,2 - 10 µl-32 mmPink10 racks, 96 each960Log in for availabilityLog in for availability1 * 960 items1 * 960 items - 91,90 €91,90 €
Sale Valid until 30/06/2025
Sale Valid until 30/06/2025New Customer Promotional Price1 * 960 items91,90 €Specifications:Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 612-25089400327040,2 - 10 µl-32 mmPink20×192/refill3.840Log in for availabilityLog in for availability1 * 3.840 items1 * 3.840 items - 296,00 €296,00 €
Sale Valid until 30/06/2025
Sale Valid until 30/06/2025New Customer Promotional Price1 * 3.840 items296,00 €Specifications:Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 613-26019400310010,2 - 10 µl-32 mmBulk1.000Log in for availabilityLog in for availability1 * 1.000 items1 * 1.000 items - 70,90 €70,90 €
Sale Valid until 30/06/2025
Sale Valid until 30/06/2025New Customer Promotional Price1 * 1.000 items70,90 €Specifications:Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 613-26009400303030,2 - 10 µl+32 mmPink10 racks, 96 each960Log in for availabilityLog in for availability1 * 960 items1 * 960 items - 116,00 €116,00 €
Sale Valid until 30/06/2025
Sale Valid until 30/06/2025New Customer Promotional Price1 * 960 items116,00 €Specifications:Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact
Finntip™ 20, colour code: Violet
Product Information
- Catalog #Supplier #SeqVolumeSterileLengthColour codePackedPkAvailabilityPrice
- 613-26069400620060,2 - 20 µl-33 mmBulk1.000Log in for availabilityLog in for availability1 * 1.000 items1 * 1.000 items - 85,30 €85,30 €
Sale Valid until 30/06/2025
Sale Valid until 30/06/2025New Customer Promotional Price1 * 1.000 items85,30 €Specifications:Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 613-26049400610070,2 - 20 µl-33 mmViolet10 racks, 384 each3.840Log in for availabilityLog in for availability1 * 3.840 items1 * 3.840 items - 554,00 €554,00 €
Sale Valid until 30/06/2025
Sale Valid until 30/06/2025New Customer Promotional Price1 * 3.840 items554,00 €Specifications:Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 613-26059400613080,2 - 20 µl+33 mmViolet10 racks, 384 each3.840Log in for availabilityLog in for availability1 * 3.840 items1 * 3.840 items - 722,00 €722,00 €
Sale Valid until 30/06/2025
Sale Valid until 30/06/2025New Customer Promotional Price1 * 3.840 items722,00 €Specifications:Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact
Finntip™ 50, colour code: Turquoise
Product Information
- Catalog #Supplier #SeqVolumeSterileLengthColour codePackedPkAvailabilityPrice
- 612-63539400373110,5 - 50 µl+48 mmTurquoise10 racks, 384 each3.840Log in for availabilityLog in for availability1 * 3.840 items1 * 3.840 items - 765,00 €765,00 €
Sale Valid until 30/06/2025
Sale Valid until 30/06/2025New Customer Promotional Price1 * 3.840 items765,00 €Specifications:Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 613-26039400360090,5 - 50 µl-48 mmBulk1.000Log in for availabilityLog in for availability1 * 1.000 items1 * 1.000 items - 86,80 €86,80 €
Sale Valid until 30/06/2025
Sale Valid until 30/06/2025New Customer Promotional Price1 * 1.000 items86,80 €Specifications:Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 612-63469400370100,2 - 50 µl-48 mmTurquoise10 racks, 384 each3.840Log in for availabilityLog in for availability1 * 3.840 items1 * 3.840 items - 623,00 €623,00 €
Sale Valid until 30/06/2025
Sale Valid until 30/06/2025New Customer Promotional Price1 * 3.840 items623,00 €Specifications:Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact
Finntip™ 250 Universal, colour code: Yellow
Product Information
- Catalog #Supplier #SeqVolumeSterileLengthColour codePackedPkAvailabilityPrice
- 613-25959400230130,5 - 250 µl-52 mmBulk1.000Log in for availabilityLog in for availability1 * 1.000 items1 * 1.000 items - 63,80 €63,80 €
Sale Valid until 30/06/2025
Sale Valid until 30/06/2025New Customer Promotional Price1 * 1.000 items63,80 €Specifications:Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 612-63439400267170,5 - 250 µl-52 mmYellow20×96/refill1.920Log in for availabilityLog in for availability1 * 1.920 items1 * 1.920 items - 143,00 €143,00 €
Sale Valid until 30/06/2025
Sale Valid until 30/06/2025New Customer Promotional Price1 * 1.920 items143,00 €Specifications:Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 613-25979400260150,5 - 250 µl-52 mmYellow10 racks, 96 each960Log in for availabilityLog in for availability1 * 960 items1 * 960 items - 82,50 €82,50 €
Sale Valid until 30/06/2025
Sale Valid until 30/06/2025New Customer Promotional Price1 * 960 items82,50 €Specifications:Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 613-25989400263160,5 - 250 µl+52 mmYellow10 racks, 96 each960Log in for availabilityLog in for availability1 * 960 items1 * 960 items - 105,00 €105,00 €
Sale Valid until 30/06/2025
Sale Valid until 30/06/2025New Customer Promotional Price1 * 960 items105,00 €Specifications:Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact
Finntip™ 300, colour code: Orange
Product Information
- Catalog #Supplier #SeqVolumeSterileLengthColour codePackedPkAvailabilityPrice
- 613-26159401260205 - 300 µl-52 mmBulk1.000Log in for availabilityLog in for availability1 * 1.000 items1 * 1.000 items - 64,30 €64,30 €
Sale Valid until 30/06/2025
Sale Valid until 30/06/2025New Customer Promotional Price1 * 1.000 items64,30 €Specifications:Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 613-26149401255235 - 300 µl-52 mmOrange10×192/refill1.920Log in for availabilityLog in for availability1 * 1.920 items1 * 1.920 items - 154,00 €154,00 €
Sale Valid until 30/06/2025
Sale Valid until 30/06/2025New Customer Promotional Price1 * 1.920 items154,00 €Specifications:Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 613-26129401250215 - 300 µl-52 mmOrange10 racks, 96 each960Log in for availabilityLog in for availability1 * 960 items1 * 960 items - 83,10 €83,10 €
Sale Valid until 30/06/2025
Sale Valid until 30/06/2025New Customer Promotional Price1 * 960 items83,10 €Specifications:Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 613-26139401253225 - 300 µl+52 mmOrange10 racks, 96 each960Log in for availabilityLog in for availability1 * 960 items1 * 960 items - 106,00 €106,00 €
Sale Valid until 30/06/2025
Sale Valid until 30/06/2025New Customer Promotional Price1 * 960 items106,00 €Specifications:Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact
Finntip™ 1000, colour code: Blue
Product Information
- Catalog #Supplier #SeqVolumeSterileLengthColour codePackedPkAvailabilityPrice
- 613-4484940111328100 - 1000 µl+71 mmBlue10 racks, 96 each960Log in for availabilityLog in for availability1 * 960 items1 * 960 items - 114,00 €114,00 €
Sale Valid until 30/06/2025
Sale Valid until 30/06/2025New Customer Promotional Price1 * 960 items114,00 €Specifications:Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 613-2608940103025100 - 1000 µl-71 mmBulk1.000Log in for availabilityLog in for availability1 * 1.000 items1 * 1.000 items - 64,60 €64,60 €
Sale Valid until 30/06/2025
Sale Valid until 30/06/2025New Customer Promotional Price1 * 1.000 items64,60 €Specifications:Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 613-4485940111529100 - 1000 µl-71 mmBlue10×96/refill960Log in for availabilityLog in for availability1 * 960 items1 * 960 items - 81,10 €81,10 €
Sale Valid until 30/06/2025
Sale Valid until 30/06/2025New Customer Promotional Price1 * 960 items81,10 €Specifications:Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 612-6378940111027100 - 1000 µl-71 mmBlue10 racks, 96 each960Log in for availabilityLog in for availability1 * 960 items1 * 960 items - 88,40 €88,40 €
Sale Valid until 30/06/2025
Sale Valid until 30/06/2025New Customer Promotional Price1 * 960 items88,40 €Specifications:Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact
Finntip™ 5 ml, colour code: Green
Product Information
- Catalog #Supplier #SeqVolumeSterileLengthColour codePackedPkAvailabilityPrice
- 613-5009940207033500 - 5000 µl-147 mmGreen5 racks, 54 each270Log in for availabilityLog in for availability1 * 270 items1 * 270 items - 85,60 €85,60 €
Sale Valid until 30/06/2025
Sale Valid until 30/06/2025New Customer Promotional Price1 * 270 items85,60 €Specifications:Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 613-2622940203031500 - 5000 µl-147 mmBulk500Log in for availabilityLog in for availability1 * 500 items1 * 500 items - 101,00 €101,00 €
Sale Valid until 30/06/2025
Sale Valid until 30/06/2025New Customer Promotional Price1 * 500 items101,00 €Specifications:Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 613-5010940207334500 - 5000 µl+147 mmGreen5 racks, 54 each270Log in for availabilityLog in for availability1 * 270 items1 * 270 items - 109,00 €109,00 €
Sale Valid until 30/06/2025
Sale Valid until 30/06/2025New Customer Promotional Price1 * 270 items109,00 €Specifications:Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact
Finntip™ 10 ml, colour code: Red
Product Information
- Catalog #Supplier #SeqVolumeSterileLengthColour codePackedPkAvailabilityPrice
- 613-74389402180371000 - 10000 µl-150 mmRed5 racks, 24 each120Log in for availabilityLog in for availability1 * 120 items1 * 120 items - 58,10 €58,10 €
Sale Valid until 30/06/2025
Sale Valid until 30/06/2025New Customer Promotional Price1 * 120 items58,10 €Specifications:Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 613-74399402183381000 - 10000 µl+150 mmRed5 racks, 24 each120Log in for availabilityLog in for availability1 * 120 items1 * 120 items - 72,60 €72,60 €
Sale Valid until 30/06/2025
Sale Valid until 30/06/2025New Customer Promotional Price1 * 120 items72,60 €Specifications:Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact - 613-74379402171351000 - 10000 µl-150 mmRedBulk100Log in for availabilityLog in for availability1 * 100 items1 * 100 items - 22,80 €22,80 €
Sale Valid until 30/06/2025
Sale Valid until 30/06/2025New Customer Promotional Price1 * 100 items22,80 €Specifications:Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Certificate Registration No. EMS 623400)Low Manufacturing Impact