You Searched For: Pipette Controllers
51  results were found
SearchResultCount:"51"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Pipette controller for glass and plastic pipettes from 1 to 100 ml.
Supplier: VWR Collection
Description: Pipette controller, Pro, US-plug
Catalog Number: (612-4552)
Supplier: VWR Collection
Description: Pipette controller for glass and plastic pipettes from 1 to 100 ml.
UOM: 1 * 1 items
Catalog Number: (612-4546)
Supplier: VWR Collection
Description: Pipette filler for glass and plastic pipettes from 1 to 25 ml.
UOM: 1 * 1 items
Supplier: VWR Collection
Description: The Smoothieâ„¢ pipette filler is a simple manual pipetting aid for convenient single-handed operations. It fits all sizes of glass or plastic pipettes ...
Catalog Number: (612-4548)
Supplier: VWR Collection
Description: Pipette filler for glass and plastic pipettes from 1 to 100 ml.
UOM: 1 * 1 items
Catalog Number: (612-7179)
Supplier: VWR Collection
Description: The VWR® pipette controller is a compact and portable device that features multiple user-selectable modes and speeds to perform a wide range of liquid...
UOM: 1 * 1 items
New Product
Catalog Number: (612-1912)
Supplier: Brand
Description: Suitable for disposable micropipettes with ring mark and other small volume pipettes up to 1 ml (e.g. blood diluting pipettes) with aspiration end dia...
UOM: 1 * 1 items
Supplier: Brand
Description: The accu-jet® S pipette controller brings versatility and endurance to your team. It makes lab work simple, easy, and efficient, whether you work with...
Low Manufacturing Impact
Sustainable Packaging
Supplier: Brand
Description: Macro pipette controller for glass and plastic pipettes from 0,1 to 200 ml.
Catalog Number: (612-1910)
Supplier: Brand
Description: Pipette filler for disposable micro pipettes with ring marks and small volume pipettes up to 1 ml, Ø at end <5 mm.
UOM: 1 * 1 items
Supplier: Integra
Description: Pipette controller for glass and plastic pipettes from 1 to 100 ml.
Catalog Number: (612-2715)
Supplier: Sartorius
Description: Rechargeable, cordless and lightweight, the Midi Plusâ„¢ pipette controller can be used with all plastic, glass and Pasteur-type pipettes between 1 and ...
UOM: 1 * 1 items
Supplier: Integra
Description: Pipette controller for glass and plastic pipettes from 0,1 to 100 ml.
Catalog Number: (612-1874)
Supplier: HIRSCHMANN
Description: Pipette controller for glass and plastic pipettes from 0,1 to 200 ml.
UOM: 1 * 1 items
Supplier: Bel-Art Products, a Part of SP
Description: Pipette filler for glass and plastic pipettes up to 0,2, 2, 10 or 25 ml.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 51
no targeter for Bottom