You Searched For: Microcapillaries
34  results were found
SearchResultCount:"34"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Schott AG
Description: High precision capillaries made from borosilicate 3.3 glass.
Supplier: HIRSCHMANN
Description: Disposable capillary pipettes with the end-to-end system. With genuine Schott DURAN® glass. Accuracy ±≤0,5%, CV ±≤1%. With batch number on the package...
Supplier: HIRSCHMANN
Description: Clear glass, both end open.
Supplier: HIRSCHMANN
Description: DURAN®, borosilicate glass, not heparinised, disposable.
Supplier: Vitrex Medical A/S
Description: For single use.
Supplier: HIRSCHMANN
Description: Openend®, disposable capillary pipettes, sodium heparinized, DURAN®, certified conformity, with predefined breaking point for exact volume configurati...
Supplier: Vitrex Medical A/S
Description: These tubes are supplied fire polished at the colour-coded end, which should be sealed with the Sigillum Wax. Fire polishing reduces wear on the centr...
Supplier: HIRSCHMANN
Description: Disposable capillaries for centrifugation of blood tests.
Supplier: HIRSCHMANN
Description: DURAN®, borosilicate glass, with ring mark, disposable.
Supplier: Vitrex Medical A/S
Description: Vitrex® pre-calibrated pipettes are for users who require very accurate capillary tube volume. The pipettes are manufactured to ISO 7550.
Supplier: Vitrex Medical A/S
Description: These disposable capillary tubes for blood gas analyses are made from heavy walled glass.
Supplier: HIRSCHMANN
Description: DURAN®, borosilicate glass, sodium-heparinised, disposable.
Catalog Number: (MODU569013)
Supplier: Vitrex Medical A/S
Description: This capillary dispenser is a unique liquid handling tool, developed to empty capillary tubes for blood or other liquids.
UOM: 1 * 1 items
Supplier: Brand
Description: Konformitätsbescheinigt.
Supplier: MARIENFELD
Description: These micro pipettes made of glass can dispense more accurately than conventional pipettes. They are used for dispensing very small amounts of liquids...
Supplier: GLASWARENFABRIK KARL HECHT
Description: Sodium heparinized micro haematocrit capillaries with or without mark.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 34
no targeter for Bottom