Graduated pipettes, SILBERBRAND ETERNA, type 3, Class B, total delivery
Supplier: Brand
SILBERBRAND ETERNA
612-1153EA 5.9 EUR
612-1153 612-1154 612-1155 612-1145 612-1156 612-1152 612-1147 612-1148 612-1149
Graduated pipettes, SILBERBRAND ETERNA, type 3, Class B, total delivery
Pipettes Graduated Pipettes
AR-Glas®.
  • Calibrated to deliver (TD, Ex)
  • Marks and inscriptions in ETERNA amber stain
  • Marked pipettes (*) suitable for cotton plugs
DIN EN ISO 835
Length: 360 ±10 mm except for 25 ml pipettes: 450 ±10 mm.
Certifikace: DIN EN ISO 835.