You Searched For: NS Joint Sleeves and Connectors
15  results were found
SearchResultCount:"15"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: PTFE, 0,05 mm thick.
Supplier: VWR Collection
Description: PTFE, with grip.
Supplier: Bohlender
Description: Spherical ground joint sleeves, made of PTFE, with gripping rings and sealing rings on the outside; seal suitable for high vacuums. Robust design for ...
Supplier: Bohlender
Description: PTFE.
Supplier: Bohlender
Description: Reusable sleevesmade of PTFE, with gripping ring and sealing rings on the outside.
Supplier: SciLabware
Description: Sleeves, PTFE, for conical ground joints.
Supplier: COWIE
Description: Made of PTFE, grease-free, high vacuum full length rigid sleeves for standard glass joints, with substantial shoulder for ease of handling and repeate...
Supplier: SciLabware
Description: Tapered sleeves, made of PTFE, for grease free and high vacuum seals using ground glass joints.
Supplier: COWIE
Description: Thin wall joint sleeves, made of PFTE, are inserted between standard taper ground glass joints to eliminate the use of grease and prevent sticking.
Supplier: KARTELL
Description: PTFE.
Catalog Number: (BRND51422)
Supplier: Brand
Description: PTFE, 0,05 mm thick.
UOM: 1 * 10 items
Supplier: GLASWARENFABRIK KARL HECHT
Description: Disposable sleeves for ground joints and stoppers, prevent the ground joints from sticking together.
Catalog Number: (SEMA0614)
Supplier: SEMADENI
Description: Made of PTFE, white.
UOM: 1 * 1 items
Supplier: Lenz Laborglas GmbH & CO.KG
Description: Sleeves, made of PTFE. For fitting onto standard ground cones.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: Disposable ground joint sleeves. thin walled, made of PTFE.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 15 of 15
no targeter for Bottom