You Searched For: Mortars and Pestles
39  results were found
SearchResultCount:"39"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Agate.
Supplier: VWR Collection
Description: Agate.
Supplier: VWR Collection
Description: Agate.
Supplier: VWR Collection
Description: Porcelain.
Supplier: VWR Collection
Description: Porcelain, produced according to DIN 12906.
Supplier: VWR Collection
Description: Wear-proof purple polypropylene pestles which are designed to fit into cell strainers perfectly.
Supplier: BOCHEM
Description: Stainless steel 18/10.
Catalog Number: (BOCH8936)
Supplier: BOCHEM
Description: 18/10 Stainless steel.
UOM: 1 * 1 items
Supplier: Retsch GmbH
Description: Agate. High quality with flawless grinding surface.
Supplier: USBECK Laborgeräte
Description: Mortar and pestle made of stainless steel.
Catalog Number: (AXYGPES-15-B-SI)
Supplier: Corning
Description: Durable and inert polypropylene construction
UOM: 1 * 400 items
Supplier: BOHEMIA CRISTAL HANDELSGES.MBH
Description: Mortar with Pestle, Height: 90 mm
Supplier: GLASWARENFABRIK KARL HECHT
Description: Borosilicate glass, with spout and rough interior surface.
Catalog Number: (NUNC533669)
Supplier: Thermo Fisher Scientific
Description: Mortar with Pestle, Sterile kit, Nuncâ„¢, HDPE
UOM: 1 * 50 items
Supplier: VITLAB
Description: White Melamine, with lip.
Supplier: GLASWARENFABRIK KARL HECHT
Description: Porcelain, with spout.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 39
no targeter for Bottom