PCR tube rack
732-0810EA 51.8 EUR
VWRI732-0810
PCR tube rack
PP.
  • Separate removable 96-well rack holds a plate, 0,2 ml tubes or 0,2 ml tube strips
  • Additional wells on the workstation accommodate 0,5; 1,5 or 2,0 ml tubes
  • Autoclavable and freezable
  • Sustainable design based on virgin resins
Suitable for preparing samples for cycling, or working with completed procedures.