Full face masks, 7907S
Supplier: 3M
111-0094EA 384 EUR
MMMA7907S
Full face masks, 7907S
Lightweight full face masks for maximum comfort. The face piece is manufactured from a soft hypo-allergenic material and incorporates a central exhalation valve. Speech diaphragm for improved communications.
  • Uses 6000/2000 series bayonet filters
  • Large lens, with a scratch resistant coating gives a wide field of vision and excellent visibility
  • Dual filter design reduces breathing resistance and improves weight distribution for greater comfort
  • Double face sealing gasket ensures a better fit
  • Six strap harness assembly for easy adjustment and reliable fit
Certifikace: EN 136
Informace pro objednávání: Supplied without filters, which are sold separately.
Upozornení: *One size fits all