Aluminium, anodised in yellow/black. Height-adjustable work surfaces for precise placement of tools and containers on laboratory apparatus. Very stable model due to special two-spindle system.
- Available with or without casters with fixed or free-running capability
- Two-spindle system assures exceptional stability
- Capable of sustaining heavy loads
- Corrosion resistant and resistant to a wide range of chemicals
- Remote controllable
On models with an electric motor, the desired bottom and top limit positions can be adjusted to any position. When it reaches this position, the lifting platform stops automatically. These lifting platforms are also suitable for repetitive work steps. They can be controlled remotely and their speed can be controlled steplessly.