You Searched For: Labels
588  results were found
SearchResultCount:"588"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: These labels, made of PE film and with the GHS symbols, meet with GHS (Global Harmonised System) regulations for chemical substances and mixtures. The...
Supplier: VWR Collection
Description: The labels and adhesive have been designed to withstand temperatures from –196 to +121 °C. VWR® labels are ideal for all cryostorage, and can also be ...
Supplier: VWR Collection
Description: PolarSafe Dots are made from chemically inert polyester with a unique adhesive formulated to adhere to laboratory glass and plastic ware. These dots w...
Supplier: VWR Collection
Description: These label dot rolls are made from chemically-inert polyester with a unique adhesive formulated to adhere to laboratory glass and plasticware.
Supplier: Brady
Description: These BradyJet J2000 SFID printer labels are used to produce eye-catching safety signs, tags, pipe markers and lean instruction labels to increase wor...
Supplier: Brady
Description: The B-494 polyester label is a reliable sample identification and traceability solution.
Supplier: Brady
Description: Static dissipative polyimide (B-718) withstands wave solder environments and multiple harsh washes for printed circuit board and electronic component ...
New Product
Catalog Number: (312-0103)
Supplier: BÃœRKLE
Description: These safety labels are used as originality seals and as protection from manipulations of e.g. samples, retention samples, for transporting samples, s...
UOM: 1 * 1 Roll
Catalog Number: (BRDY173488)
Supplier: Brady
Description: Durable FreezerBondzâ„¢ Polyester (B-492) markers are designed for laboratory identification applications such as vials, centrifuge, tubes, test tubes, ...
UOM: 1 * 1 Roll
New Product
Catalog Number: (SCOT294010203)
Supplier: DWK Life Sciences
Description: These self-adhesive, white polyester labels are designed for use with the DURANâ„¢ YOUTILITY bottle system.
UOM: 1 * 1 items
Supplier: Brady
Description: These inspection date labels are made of vinyl cloth. They can be used wherever a frequent and regular inspection of machinery and equipment is requir...
Supplier: Brady
Description: These water dissolvable paper labels are ideal to label items that need new identification after every use. Exposing the label to warm water will comp...
Catalog Number: (BRDY016104)
Supplier: Brady
Description: Sized to fit popular dot-matrix, thermal transfer and laser label sizes. Liners are backslit for easy overlaminate removal.
UOM: 1 * 1 Roll
Catalog Number: (479-3291)
Supplier: Thermo Fisher Scientific
Description: Cloth labels for use at ultra-low temperatures, ballpoint pen compatible.
UOM: 1 * 200 items
Supplier: Brady
Description: Easy-to-use vinyl labels can help to cut material waste, save label making time, and print crisp labels very efficiently.
Supplier: Brady
Description: These glycol energy source tags are constructed of PP with round corners with one hole.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 588
no targeter for Bottom