You Searched For: Knives
39  results were found
SearchResultCount:"39"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: SWANN-MORTON
Description: Heavy duty trimming knifes with fixed or retractable blade.
Supplier: RSG
Description: These ceramic laboratory knives have extremely hard blades which resist chemicals, fractures and corrosion.
Supplier: RSG
Description: Stainless steel blade, plastic handle.
Supplier: RSG
Description: Chromium molybdenum vanadium steel blade, POM handle.
Supplier: SWANN-MORTON
Description: Heavy-duty cast metal handles with either fixed blade or retractable blade mechanism. Ideal for industrial applications as well as for D.I.Y. enthusia...
Catalog Number: (RSGA192.034)
Supplier: RSG
Description: This steak knife has a special grinding in the front area of the blade. The blade is made of stainless chromium steel, the handle is made of POM plast...
UOM: 1 * 1 items
Supplier: SWANN-MORTON
Description: These knives with nylon handle are particularly suitable for artwork, crafts and model making.
Catalog Number: (RSGA181.099)
Supplier: RSG
Description: These knives are ideal for chopping and cutting fish, meat, vegetables and fruits. The blade is made of chromium steel, the handle is made of POM plas...
UOM: 1 * 1 items
Catalog Number: (RSGA190.004)
Supplier: RSG
Description: This knife set includes one larding knife, two ham slicers (different sizes), and one cooking knife. Packed in a black case.
UOM: 1 * 1 SET
Catalog Number: (233-1725)
Supplier: RSG
Description: Stainless steel blade, plastic handle.
UOM: 1 * 1 items
Catalog Number: (233-0193)
Supplier: MARTOR ARGENTAX
Description: Steel blade, handle made of glass fibre reinforced plastic.
UOM: 1 * 1 items
Catalog Number: (217-9693)
Supplier: USBECK Laborgeräte
Description: Metal blade, plastic handle.
UOM: 1 * 1 items
Catalog Number: (233-0196)
Supplier: MARTOR ARGENTAX
Description: Trapezoid blade, handle made of solid die-cast zinc.
UOM: 1 * 1 items
Catalog Number: (223-0099)
Supplier: MARTOR ARGENTAX
Description: Steel blade, aluminium handle.
UOM: 1 * 1 items
Catalog Number: (BOCH12230)
Supplier: BOCHEM
Description: Jack-knife, collapsible, with stainless steel blade, wooden handle and hook for hanging up.
UOM: 1 * 1 items
Supplier: Sampling Systems
Description: Knife, SteriWare®, HDPE, Colour: White, Blade: 60 mm, Total length: 160 mm
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 39
no targeter for Bottom