You Searched For: Spirit Lamps
8  results were found
SearchResultCount:"8"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (451-3120)
Supplier: USBECK Laborgeräte
Description: Stainless steel.
UOM: 1 * 1 items
Catalog Number: (HECH43031010)
Supplier: GLASWARENFABRIK KARL HECHT
Description: Glass alcohol lamps without lateral tubing, complete with wick and brass wick holder.
UOM: 1 * 1 items
Supplier: DWK Life Sciences
Description: Soda-lime glass with ground glass joint cap. The spirit lamp is an alternative to the gas-powered Bunsen burner and is also suitable for heating cooki...
Catalog Number: (WHEA237070)
Supplier: DWK Life Sciences
Description: Alcohol burner manufactured from soda-lime glass, designed for use with isopropyl or denatured ethyl alcohol.
UOM: 1 * 6 items
Catalog Number: (451-0096)
Supplier: BOCHEM
Description: Spirit lamp made of 18/10 stainless steel.
UOM: 1 * 1 items
Catalog Number: (HERE1082601)
Supplier: HERENZ, HEINZ GMBH
Description: Heater, Spirit lamp, Spirit lamp, with wick and spout, Glass, 100 ml
UOM: 1 * 1 items
Catalog Number: (RSGA253.000)
Supplier: RSG
Description: Heater, Spirit lamp, Burner, with wick regulation and cowl, Stainless steel, ±60 ml
UOM: 1 * 1 items
Catalog Number: (MURRG671/02)
Supplier: MURRAY, S & CO LTD
Description: Heater, Spirit lamp, Spirit lamp, glass with fitted wick in ceramic burner, Glass, 100 ml
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8 of 8
no targeter for Bottom