You Searched For: Gas Tubes
45  results were found
SearchResultCount:"45"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: ROBU GLASFILTER
Description: These versatile and recyclable tubes offer excellent properties for diverse applications are made of borosilicate glass 3.3.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: Gas distribution tubes with lateral ring made of borosilicate glass 3,3.
Supplier: GASTEC
Description: The dositubes are used to measure time-weighted average (TWA) gas concentrations over longer periods.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: Gas distribution tubes with central ring made of borosilicate glass 3.3.
Supplier: GASTEC
Description: Crystal clear gas tubes with distinct colour changes.
Supplier: DWK Life Sciences
Description: These gas distribution tubes are designed for reactions between gases and liquids. They are made of clear DURAN® borosilicate glass 3.3.
Supplier: Dräger
Description: To measure the 'spot concentrations' of water vapour for use with an Accuro® pump.
Supplier: Dräger
Description: To measure the 'spot concentrations' of carbon monoxide for use with an Accuro pump. Uses include determination and measurement of concentration peaks...
Catalog Number: (151-0145)
Supplier: Dräger
Description: Operating conditions: 0 to 40 °C ; 3 to 15 mg/l H₂O absolute humidity.
UOM: 1 * 10 items
Supplier: Dräger
Description: To measure the 'spot concentrations' of hydrochloric acid in the air with an Accuro® pump - Uses include determination and measurement of concentratio...
Supplier: Dräger
Description: To measure the 'spot concentrations' of carbon dioxide for use with an Accuro® pump. Uses include determination and measurement of concentration peaks...
Supplier: Dräger
Description: To measure the 'spot concentrations' of perchloroethylene for use with an Accuro pump. Uses include determination and measurement of concentration pea...
Supplier: Dräger
Description: To measure the 'spot concentrations' of hydrogen sulphide for use with an Accuro® pump. Uses include determination and measurement of concentration pe...
Supplier: Dräger
Description: Sampling activated tubes for direct measurement indication even with complex compounds and mixtures of substances in air investigations and gas analys...
Supplier: Dräger
Description: To measure the 'spot concentrations' of hydrocarbons with an Accuro® pump.
Supplier: Dräger
Description: Different gases and vapours can be detected and measured such as concentration peaks, personal exposure in the inhalation area, detection of leaks in ...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 45
no targeter for Bottom