Analytical funnels
Supplier: SciLabware
221-0009EA 25.85 EUR
221-0009 221-0011
Analytical funnels
Funnels Filtration Funnels Analytical Funnels
PYREX®, borosilicate glass.
  • Funnel fluted to promote rapid filtration
  • Very good chemical resistance
  • High temperature resistance