You Searched For: Funnels
317  results were found
SearchResultCount:"317"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Borosilicate 3.3 glass.
Supplier: VWR Collection
Description: PP, translucent.
Supplier: VWR Collection
Description: Borosilicate 3.3 glass.
Supplier: VWR Collection
Description: Borosilicate 3.3 glass with NS-PE stopper.
Supplier: VWR Collection
Description: Borosilicate 3.3 glass, with NS-PE stopper.
Supplier: VWR Collection
Description: Borosilicate 3.3 glass.
Sale
Supplier: VWR Collection
Description: Borosilicate 3.3 glass.
Catalog Number: (720-1999)
Supplier: VWR Collection
Description: Single-use plastic chamber used in cytocentrifugation for urine, gynaecological and non gynaecological specimens, also designed to perform cell suspen...
UOM: 1 * 600 items
Supplier: ROETZMEIER SICHERH. BEHAELTER
Description: Stainless steel.
Supplier: KARTELL
Description: 60° angle, autoclavable funnels. Large bore avoids bridging of powders during transfer. Ribbed outside to avoid pressure lock. Can be used with liquid...
Supplier: Simport Scientific
Description: These funnels will snap quickly in place, allowing the pad to align correctly with the sample tunnel. They are made with a compression ring around the...
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass, clear, with PTFE key.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass, with NS-PE stopper.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass, with NS-PE stopper.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: Separating funnels, Squibb, stopcock with glass or PTFE key, made of borosilicate glass amberized.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass, clear, with glass key.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 317
no targeter for Bottom