You Searched For: Rotary Evaporator Flasks
19  results were found
SearchResultCount:"19"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Borosilicate 3.3 glass.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass, clear.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass, clear or plastic-coated.
Supplier: Heidolph Instruments GmbH & Co.KG
Description: Evaporating flasks, Borosilicate glass, uncoated, Capacity: 1000 ml, Socket size: 24/29 NS
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass, clear or plastic-coated.
Supplier: Heidolph Instruments GmbH & Co.KG
Description: Evaporating flasks, Borosilicate glass 3.3, with flange, Capacity: 6000 ml
Supplier: DWK Life Sciences
Description: DURAN®, borosilicate glass 3.3, clear.
Supplier: DWK Life Sciences
Description: DURAN®, borosilicate glass 3.3, clear.
Catalog Number: (IKAA1905700)
Supplier: IKA
Description: Borosilicate glass 3.3.
UOM: 1 * 1 items
Supplier: GLASWARENFABRIK KARL HECHT
Description: Borosilicate glass 3.3.
Supplier: WITEG LABORTECHNIK GMBH
Description: Borosilicate glass 3.3.
Supplier: BUCHI
Description: Borosilicate glass 3.3.
Supplier: BUCHI
Description: Made of borosilicate glass.
Supplier: WITEG LABORTECHNIK GMBH
Description: Borosilicate glass 3.3.
Supplier: WITEG LABORTECHNIK GMBH
Description: Borosilicate glass, clear.
Supplier: BUCHI
Description: Made of Borosilicate glass.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 19
no targeter for Bottom