Distillation flasks, Engler type, DURAN®
Supplier: DWK Life Sciences
214-0188EA 0 EUR
SCOT212433605
Distillation flasks, Engler type, DURAN®
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With white labeling area.