You Searched For: Culture Flasks
29  results were found
SearchResultCount:"29"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: DWK Life Sciences
Description: DURAN®, borosilicate glass 3.3, clear.
Supplier: DWK Life Sciences
Description: DURAN®, borosilicate glass 3.3, clear.
Catalog Number: (391-0370)
Supplier: DWK Life Sciences
Description: This penicillin flask is made from clear DURAN® borosilicate glass 3.3.
UOM: 1 * 1 items
Catalog Number: (391-0310)
Supplier: DWK Life Sciences
Description: DURAN®, borosilicate glass 3.3, clear.
UOM: 1 * 1 items
Supplier: DWK Life Sciences
Description: DURAN®, borosilicate glass 3.3, clear.
Catalog Number: (391-0340)
Supplier: DWK Life Sciences
Description: DURAN®, borosilicate glass 3.3, clear.
UOM: 1 * 1 items
Supplier: DWK Life Sciences
Description: These baffled culture shake flasks are designed for use with rotary and reciprocating shakers where baffle indents provide greater turbulence to impro...
Catalog Number: (734-5057)
Supplier: Thermo Fisher Scientific
Description: Clear, graduated PC vessel, white PP screw caps, PTFE stirring bar and PP/TFE stirring assembly.
UOM: 1 * 1 items
Catalog Number: (734-5055)
Supplier: Thermo Fisher Scientific
Description: PC culture vessel with white PP closures.
UOM: 1 * 1 items
Catalog Number: (710-4100)
Supplier: Thermo Fisher Scientific
Description: Clear PC.
UOM: 1 * 1 items
Supplier: Bohlender
Description: Culture bottles, made of borosilicate glass, suitable for both low and high speed (maximum 1000 rpm) stirrers for gentle mixing of cell cultures.
Catalog Number: (734-0880)
Supplier: Cole-Parmer
Description: Culture flasks, borosilicate glass, siliconised, for use with the MCS platform biological stirrers, ideal for suspension cell culturing with reduced c...
UOM: 1 * 1 items
Supplier: WITEG LABORTECHNIK GMBH
Description: Culture flask, made of borosilicate glass, Erlenmeyer form, designed for orbital shakers.
Supplier: WITEG LABORTECHNIK GMBH
Description: Culture flask, made of borosilicate glass, Erlenmeyer shape, with straight neck for metal caps.
Supplier: WITEG LABORTECHNIK GMBH
Description: Culture flasks, made of borosilicate glass, Erlenmeyer shape, designed for orbital shakers.
Supplier: DWK Life Sciences
Description: These culture shake flasks are made of 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E-438 Type I, Class A requirements.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 29
no targeter for Bottom