Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (5)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
First Aid Kits
https://si.vwr.com/stibo/smallweb/std.lang.all/69/01/20236901.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"22"
  List View Easy View (new)
Sort by:
Catalog Number: (118-0395)
Supplier:  ORKLA CARE AB
Description:   This portable first aid kit contains first aid products to be used in emergency situations (bleeding, breathing, burns, sprains, circulatory shock). I...
UOM:  1 * 1 items
Catalog Number: (118-0393)
Supplier:  ORKLA CARE AB
Description:   This portable first aid kit is designed for personal use. It contains first aid products to be used in emergency situations (bleeding). Ideal for peop...
UOM:  1 * 1 items
Catalog Number: (118-0396)
Supplier:  ORKLA CARE AB
Description:   This extra large first aid kit contains first aid products for use in emergency situations (bleeding, breathing, burns, sprains, circulatory shock, mi...
UOM:  1 * 1 items
Catalog Number: (118-0394)
Supplier:  ORKLA CARE AB
Description:   This portable first aid kit is designed for personal use. It contains first aid products to be used in emergency situations (bleeding, breathing, burn...
UOM:  1 * 1 items
Catalog Number: (118-1420)
Supplier:  SOEHNGEN W. GMBH
Description:   Durable, lightweight and sleek first aid kit in nylon case, very space-saving.
UOM:  1 * 1 items
Catalog Number: (118-0242)
Supplier:  TRENKA
Description:   240×340×120 mm
UOM:  1 * 1 items
Catalog Number: (118-0241)
Supplier:  TRENKA
Description:   200×280×115 mm
UOM:  1 * 1 items
Catalog Number: (118-1410)
Supplier:  SOEHNGEN W. GMBH
Description:   This customised, professional first aid kit is suitable for use in laboratories and the chemical industry. It contains an eyewash fountain, cold compr...
UOM:  1 * 1 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 22
  1  2  3  Next 
A new look and a renewed commitment
Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.
Learn More Start Shopping