Order Entry
Syringe filters, ReZist™, Whatman™
Certificates
undefined
Syringe filters, ReZist™, Whatman™
Related Information
  •  
    Seq
    Ø
    Pore size
    Colour
    Inlet/outlet connections
    Sterility
    Pk
  • 514-1327
    10463713
    02
    13 mm
    0,45 µm
    Green
    Luer lock female/Mini-tip
    Non sterile
    100
    1 * 100 items
    1 * 100 items - 351,00 €
    351,00 €
    1 * 100 items
       351,00 €
    Specifications
    Environmentally Preferable
  • 514-1311
    10463500
    03
    30 mm
    0,20 µm
    White
    Luer lock female/Luer male
    Sterile
    50
    1 * 50 items
    1 * 50 items - 317,00 €
    317,00 €
    1 * 50 items
       317,00 €
    Specifications
    Environmentally Preferable
  • 514-1321
    10463503
    04
    30 mm
    0,20 µm
    White
    Luer lock female/Luer male
    Non sterile
    100
    1 * 100 items
    1 * 100 items - 457,00 €
    457,00 €
    1 * 100 items
       457,00 €
    Specifications
    Environmentally Preferable
  • 514-1320
    10463505
    05
    30 mm
    0,20 µm
    White
    Luer lock female/Luer male
    Non sterile
    500
    1 * 500 items
    1 * 500 items - 1 840,00 €
    1 840,00 €
    1 * 500 items
       1 840,00 €
    Specifications
    Environmentally Preferable
  • 514-1322
    10463513
    06
    30 mm
    0,45 µm
    Green
    Luer lock female/Luer male
    Non sterile
    100
    1 * 100 items
    1 * 100 items - 457,00 €
    457,00 €
    1 * 100 items
       457,00 €
    Specifications
    Environmentally Preferable
  • 514-1328
    10463515
    07
    30 mm
    0,45 µm
    Green
    Luer lock female/Luer male
    Non sterile
    500
    1 * 500 items
    1 * 500 items - 1 840,00 €
    1 840,00 €
    1 * 500 items
       1 840,00 €
    Specifications
    Environmentally Preferable
  • 514-1323
    10463523
    10
    30 mm
    1,00 µm
    Yellow
    Luer lock female/Luer male
    Non sterile
    100
    1 * 100 items
    1 * 100 items - 457,00 €
    457,00 €
    1 * 100 items
       457,00 €
    Specifications
    Environmentally Preferable
  • 514-1329
    10463525
    11
    30 mm
    1,00 µm
    Yellow
    Luer lock female/Luer male
    Non sterile
    500
    1 * 500 items
    1 * 500 items - 1 840,00 €
    1 840,00 €
    1 * 500 items
       1 840,00 €
    Specifications
    Environmentally Preferable
  • 514-1324
    10463533
    12
    30 mm
    5,00 µm
    Grey
    Luer lock female/Luer male
    Non sterile
    100
    1 * 100 items
    1 * 100 items - 457,00 €
    457,00 €
    1 * 100 items
       457,00 €
    Specifications
    Environmentally Preferable
  • 516-0444
    10463535
    13
    30 mm
    5,00 µm
    Grey
    Luer lock female/Luer male
    Non sterile
    500
    1 * 500 items
    1 * 500 items - 1 840,00 €
    1 840,00 €
    1 * 500 items
       1 840,00 €