You Searched For: Glass Fibre Filters
55  results were found
SearchResultCount:"55"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Binder-free glass fibre filters feature fast flow rates, high loading capacity, wide thermal tolerance and excellent precipitate retention.
Catalog Number: (515-2877)
Supplier: Whatman products (Cytiva)
Description: These borosilicate glass papers are designed for use when drying samples during moisture assessment.
UOM: 1 * 100 items
Supplier: Whatman products (Cytiva)
Description: These borosilicate glass filters are widely used as total suspended solids filters for applications testing both potable water and waste water. GF/C c...
Supplier: Whatman products (Cytiva)
Description: These borosilicate glass filters are suitable for water quality parameter measurement and air filtration applications. They can be used for high effic...
Supplier: Whatman products (Cytiva)
Description: Whatman Grade GF 9 glass microfiber filters from Cytiva's business are strengthened with an inorganic binder for environmental analysis of coarse, hea...
Sale
Supplier: Whatman products (Cytiva)
Description: QM-C 100% quartz microfiber filter paper features low trace metals and high temperature resistance. It is well-suited for heavy metal analysis of part...
Supplier: Whatman products (Cytiva)
Description: These high purity quartz microfibre filters are used in many air sampling filter applications, including sampling of acidic gases, stacks, flues and a...
Supplier: Whatman products (Cytiva)
Description: These borosilicate glass filters are particularly useful for liquid clarification and liquid scintillation counting.
Supplier: Whatman products (Cytiva)
Description: These borosilicate glass filters are ideal for measuring landfill leachate composition with a TCLP test. They can also be used for DNA binding and pur...
Supplier: Whatman products (Cytiva)
Description: Whatmanâ„¢ QM-H is a pure quartz fibre filter for use in testing acidic gases, stacks, flues and aerosols. It has low heavy metal content so it is well ...
Sale
Supplier: Whatman products (Cytiva)
Description: These borosilicate glass filters are specified for water pollution monitoring, cell harvesting, liquid scintillation counting and air pollution monito...
Supplier: Whatman products (Cytiva)
Description: These microfiber filters made from borosilicate glass are suitable for water and wastewater analysis, with extremely fine particle retention and added...
Supplier: Whatman products (Cytiva)
Description: Whatman Grade GF 92 glass microfiber filters from Cytiva are designed for prefiltration in order to protect against membrane fouling. Grade GF 92 is s...
Supplier: Cytiva (Formerly Pall Lab)
Description: Superior grade glass fibre filters for a variety of air sampling methods. Used in a variety of sample clean-up, prefiltration and analytical testing a...
Supplier: Whatman products (Cytiva)
Description: Cytiva's Whatman TCLP test filters are acid-treated with low metal content. They support TCLP testing by providing accurate information about TCLP lim...
Sale
Supplier: Whatman products (Cytiva)
Description: These 934-AHâ„¢ RTU grade filter papers have been pre-washed, dried (or ignited), cooled, desiccated and weighed, which eliminates the pre-treating step...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 55
no targeter for Bottom