You Searched For: Filter Discs
23  results were found
SearchResultCount:"23"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Avantor
Description: Discs are pre-assembled for use in preparing aqueous samples for analysis. Laminar configuration provides filtration capacity and inlet characteristic...
Supplier: ROBU GLASFILTER
Description: The unique porous structure of VitraPOR® sintered glass filters makes it flexible and versatile for many applications.
Supplier: ROBU GLASFILTER
Description: Filter discs made from DURAN®, borosilicate glass. Edges not fused and centred, surface untreated.
Supplier: ROBU GLASFILTER
Description: The unique porous structure of VitraPOR® sintered glass filters makes it flexible and versatile for many applications.
Supplier: ROBU GLASFILTER
Description: The unique porous structure of VitraPOR® sintered glass filters makes it flexible and versatile for many applications.
Supplier: ROBU GLASFILTER
Description: The unique porous structure of VitraPOR® sintered glass filters makes it flexible and versatile for many applications.
Supplier: ROBU GLASFILTER
Description: The unique porous structure of VitraPOR® sintered glass filters makes it flexible and versatile for many applications.
Supplier: ROBU GLASFILTER
Description: The unique porous structure of VitraPOR® sintered glass filters makes it flexible and versatile for many applications.
Supplier: ROBU GLASFILTER
Description: The unique porous structure of VitraPOR® sintered glass filters makes it flexible and versatile for many applications.
Supplier: ROBU GLASFILTER
Description: The unique porous structure of VitraPOR® sintered glass filters makes it flexible and versatile for many applications.
Supplier: Thermo Scientific
Description: These quartz discs are manufactured by fusing naturally occurring crystalline silica.
Supplier: DWK Life Sciences
Description: Sintered glass filters have neutral properties with unfused edge and untreated surfaces. The hard sintering process makes the material resilient.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: Glass filter discs centred with unfused rim.
Supplier: DWK Life Sciences
Description: These filter discs feature a fused glass rim.
Supplier: Whatman products (Cytiva)
Description: The polyester drain disc is binder free and has a thickness of 100 µm.
Catalog Number: (KAVA632424011309)
Supplier: BOHEMIA CRISTAL HANDELSGES.MBH
Description: Filter disc made from borosilicate glass. Sintered glass filter with ground edges.
UOM: 1 * 20 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 23
no targeter for Bottom