You Searched For: Face Shields
55  results were found
SearchResultCount:"55"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: JSP LTD
Description: A range of high performance industrial visors, fully compatible with EVOGuard® browguard systems and EVO® helmet models.
Supplier: UVEX
Description: Helmet visor made from strong PC protects against the effects of electrical arcs. It is compatible with uvex safety helmets and uvex 2H, uvex 3H and u...
Supplier: UVEX
Description: These polycarbonate and cellulose acetate visors can be attached to helmets by using a length adapter and can be locked in three positions.
Catalog Number: (MMMA5A-CP)
Supplier: 3M
Description: Acetate face shields are lightweight and non-conductive.
UOM: 1 * 1 items
Catalog Number: (JSPAAFA061-131-100)
Supplier: JSP LTD
Description: Faceshield offers protection against medium energy, high-speed particles at 120 m/s (270 mph), liquid droplets, and molten metal.
UOM: 1 * 1 items
Catalog Number: (JSPAADB280-000-400)
Supplier: JSP LTD
Description: EVOGuard®C Industrial Browguard is designed for use with EVOGuard® visors, to provide innovative solutions to protect the wearer in a multitude of hig...
UOM: 1 * 1 items
Catalog Number: (SCTS2019716)
Supplier: SCOTT SAFETY
Description: Interchangeâ„¢ Browguard IB2000 is designed with a durable nylon shell construction to provide a high protection and comfort level in industrial applica...
UOM: 1 * 10 items
Catalog Number: (JSPAAHV970-001-100)
Supplier: JSP LTD
Description: The Cough Guard can be fitted to a JSP bump cap to reduce the likelihood of exhaled bacteria/virus contacting the wearer's face. This device is not sp...
UOM: 1 * 1 items
Catalog Number: (111-1635)
Supplier: 3M
Description: This face shield consists of headgear (H8) made of thermoplastic and clear PC visor (WP96). Both parts provide high impact resistance. Ideal for use i...
UOM: 1 * 1 items
Supplier: 3M
Description: 3Mâ„¢ V4 series industrial Multisystem replacement face shields are available in multiple visor materials and color finishes.
Catalog Number: (JSPAAFN060-000-000)
Supplier: JSP LTD
Description: The Force®8 Splash Guard can be fitted to a Force®8 half mask to reduce the likelihood of exhaled bacteria/virus hitting the wearer's face.
UOM: 1 * 10 items
Catalog Number: (MMMA5E-11)
Supplier: 3M
Description: Face Shield 5E-11 is an optical class 1, shade 5, green tinted, curved polycarbonate faceshield with anti-fog and anti-scratch coating for protection ...
UOM: 1 * 1 items
Catalog Number: (MMMAV46)
Supplier: 3M
Description: Face shield holder for V4-Series face shields. This face shield holder with short peak is designed for use with 3Mâ„¢ V4-Series face shields.
UOM: 1 * 1 items
Supplier: UVEX
Description: Robust visor with integrated forehead cover, clear cellulose acetate lens and additional protective cap made from impact-resistant PS.
Supplier: JSP LTD
Description: A compact, comfortable, and lightweight faceshield with superior optical quality.
Catalog Number: (MMMAWP98)
Supplier: 3M
Description: The WP98 Series Face Shield is a high impact resistant, clear acetate faceshield designed to fit 3M hard hats. It can be used with the H8 Headgear eas...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 55
no targeter for Bottom