You Searched For: Extraction Thimbles
15  results were found
SearchResultCount:"15"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Pure cellulose cotton. Suitable for all types of extractors (Soxhlet, Büchi, Gerhardt and more).
Catalog Number: (537-0028)
Supplier: Whatman products (Cytiva)
Description: High-purity Whatman quartz thimbles from Cytiva support accurate, reproducible emission testing by providing low extractable levels and temperature re...
UOM: 1 * 10 items
Supplier: Whatman products (Cytiva)
Description: Cytiva's Whatman cellulose thimbles are manufactured from high-quality alpha cellulose cotton linter for high-performance use in automated Soxhlet ext...
Supplier: ROBU GLASFILTER
Description: Borosilicate glass 3.3.
Supplier: Whatman products (Cytiva)
Description: Cytiva's Whatman 603G borosilicate glass fiber thimbles with inorganic binder are flexibly designed for use in a full range of commercially available ...
Supplier: Whatman products (Cytiva)
Description: Grade 603 standard cellulose extraction thimbles, made of high quality cellulose, are available in a range of sizes suitable for use in thimble holder...
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, Borosilicate glass.
Supplier: Whatman products (Cytiva)
Description: Cytiva's Whatman 100% borosilicate glass microfiber (HP-GF) thimbles are designed to fit most Soxhlet extraction units. They are well-suited to pollut...
Supplier: Whatman products (Cytiva)
Description: Grade 603 standard cellulose extraction thimbles for DIONEX ASE.
Supplier: Ahlstrom-Munksjö
Description: Grade ET/401
Particularly suitable for the extraction of organic combinations out of reaction mixtures, food, natural substances, lacquer ...
Catalog Number: (537-4528)
Supplier: Ahlstrom-Munksjö
Description: Grade ET/401, cellulose.
UOM: 1 * 25 items
Supplier: MACHEREY-NAGEL
Description: Made from glass micro fibers without binder.
Supplier: MACHEREY-NAGEL
Description: Extraction thimbles, made of pure cellulose, standard quality. With or without lid available.
Supplier: BEHR
Description: Thimble, Extraction, Extraction thimble for 100 ml soxhlet extractor EZ 100 (EZ 100/H), Type: EX 100 HS
Supplier: SciLabware
Description: Borosilicate glass.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 15 of 15
no targeter for Bottom