You Searched For: Dissection Needles
34  results were found
SearchResultCount:"34"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: RSG
Description: Stainless steel.
Supplier: RSG
Description: Stainless steel needles with stainless steel handle.
Supplier: RSG
Description: Stainless steel needles with wooden handle.
Supplier: RSG
Description: Stainless steel needles with black plastic handle.
Supplier: BOCHEM
Description: Dissecting needles completely made of ¹⁸/₁₀ stainless steel.
Supplier: BOCHEM
Description: Dissecting needles made of stainless steel, with plastic handle.
Supplier: USBECK Laborgeräte
Description: Stainless steel needles with plastic handles.
Catalog Number: (HECH42712010)
Supplier: GLASWARENFABRIK KARL HECHT
Description: Dissecting needles made of chromium-nickel steel, stainless, with wooden handle.
UOM: 1 * 1 items
Supplier: USBECK Laborgeräte
Description: Stainless steel.
Supplier: HAMMACHER KARL
Description: These dissecting needles and seekers have metal handles.
Supplier: HAMMACHER KARL
Description: Dental probes made of WIRONIT steel.
Catalog Number: (233-0007)
Supplier: MURRAY, S & CO LTD
Description: Stainless steel dissecting seeker.
UOM: 1 * 10 items
Supplier: AESCULAP AG & CO
Description: Eye needles, Aesculap®, Eye needles, 3/8 circle, cutting, Fig. 3, 20 mm
Supplier: AESCULAP AG & CO
Description: 3/8 circle suture needles with round body. Heavy version.
Supplier: AESCULAP AG & CO
Description: Surgical needles, cutting, Aesculap®, Surgical needles, Fig. 6, cutting to the inner side, 1,1 mm, 32 mm
Supplier: HAMMACHER KARL
Description: These dissecting needles are made of stainless steel.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 34
no targeter for Bottom