VWR®, Disposable Weighing and Drying Dishes or Pans
Supplier: VWR
VWR®
611-1354EA 40.1 EUR
611-1354 611-1353 611-1352 611-1351 611-1350 611-3658 611-3659 611-1380
VWR®, Disposable Weighing and Drying Dishes or Pans
Boats Weighing Boats
Dishes and pans, made of aluminium.
  • Compatible with most balances and dryers
  • Oil-free
  • Slightly tapered for easy stacking
Sturdy, disposable dishes and pans. Ideal for general purpose weighing, dispensing, storage and drying applications.