Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (18)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
desiccator-lids
https://si.vwr.com/stibo/smallweb/std.lang.all/51/04/18945104.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"7"
  List View Grid View Easy View (new)
Sort by:
Supplier:  Brand
Description:   Desiccator, Lid, With socket, Novus, Nominal size: 250 DN
Supplier:  DWK Life Sciences
Description:   Plain lids, flat flange ground to fit. Lids have a knob, an NS socket or a screw thread outlet.
Supplier:  DWK Life Sciences
Description:   Desiccator, Lid, DURAN® Borosilicate glass, NS 24/29, WERTEX, Nominal size: 150 DN
Supplier:  Brand
Description:   Desiccator, Lid, DURAN® Borosilicate glass, With knob, Novus, Nominal size: 200 DN
Catalog Number: (CORN3121-250CO)
Supplier:  Corning Life Science
Description:   Desiccator, Lid, Pyrex NS 24/29, Nominal size: 250
UOM:  1 * 1 items
Supplier:  BOHEMIA CRISTAL HANDELSGES.MBH
Description:   Plastic handle, 213mm diameter edge 1 * 1 items
Supplier:  SCILABWARE
Description:   Desiccator, Lid, With socket size 24/29, for desiccators vacuum type, Nominal size: 200 DN
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 7  of 7