You Searched For: Desiccants
9  results were found
SearchResultCount:"9"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Thermo Scientific
Description: CAS No.: 81133-20-2
Supplier: Thermo Scientific
Description: CAS No.: 81133-20-2
Catalog Number: (BOHLV1895-12)
Supplier: Bohlender
Description: Silica gel with colour indicator.
UOM: 1 * 10 L
Catalog Number: (WHAT10548239)
Supplier: Whatman products (Cytiva)
Description: Desiccant pack ensure that DNA sampling cards remain dry during transport or storage.
UOM: 1 * 100 items
Supplier: Bel-Art Products, a Part of SP
Description: Convenient, easy to handle packs filled with moisture absorbing silica gel beads are available in multiple sizes to fit most desiccators.
Catalog Number: (BELAF420740020)
Supplier: Bel-Art Products, a Part of SP
Description: These colour-indicating silica gel beads are for use in refilling the Secador® auto-desiccator module which automatically regenerates them for thousan...
UOM: 1 * 25 g
Catalog Number: (STRO669039AG37)
Supplier: STROEBEL GMBH
Description: These robust, compact and versatile desiccant bags are supplied with silica gel used for pharmaceutical products and test tubes, as well as for clinic...
UOM: 1 * 1.000 items
Catalog Number: (467-4195)
Supplier: STROEBEL GMBH
Description: Protects articles from damage by humidity in the air.
UOM: 1 * 400 items
Catalog Number: (467-0000)
Supplier: Bel-Art Products, a Part of SP
Description: Reusable canister which is ideal for use in desiccator cabinets, cupboards, containers, tool boxes and other storage areas to prevent rust and moistur...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 9 of 9
no targeter for Bottom