Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Coats
https://si.vwr.com/stibo/smallweb/std.lang.all/50/49/18945049.jpg
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results.
SearchResultCount:"108"
  List View Easy View (new)
Sort by:
Supplier:  VWR Collection
Description:   The VWR Basic SPP lab coats are manufactured from a strong, lightweight, and breathable spunbonded polypropylene (SPP) fabric, which protects against ...
Supplier:  VWR Collection
Description:   These anti-static coats are manufactured from 99% polyester fibres with 1% Hydro-Tec finish (carbon treated), which makes them both extremely durable ...
Supplier:  VWR Collection
Description:   These reusable coats for women are either made of 100% cotton or 65% polyester/35% cotton. Both fabrics are robust, tear resistant and durable. The co...
Supplier:  VWR Collection
Description:   These reusable coats for women and men are made of 63% polyester, 35% cotton and 2% carbon, which provides anti-static protection. All models feature ...
Supplier:  VWR Collection
Description:   These disposable coats, with collar, are made from non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime and certain dr...
Supplier:  VWR Collection
Description:   These unisex lab coats are made from 65% polyester and 35% cotton.
Supplier:  VWR Collection
Description:   These disposable coats made of PP are ideal for visitors in non hazardous environments.
Catalog Number: (113-8265)
Supplier:  VWR Collection
Description:   This disposable smock is made from 65% LDPE and 35% LLDPE.
UOM:  1 * 100 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 108
  1  2  3  4  5  6  7  8  9  10  11  12  13  14  Next 
A new look and a renewed commitment
Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.
Learn More Start Shopping