Membrane screw caps for laboratory bottles, DURAN®
Supplier: DWK Life Sciences
215-0089EA 0 EUR
215-0089
Membrane screw caps for laboratory bottles, DURAN®
Closures Bottle Closures
PP, blue, with welded-in PTFE membrane 0,2 µm.
  • Heat resistant up to +140 °C
  • Two-way membrane with pressure equalisation of 0,1 bar
  • Ideal for autoclaving processes (does not need to be opened and closed repeatedly before/after autoclaving)
Suitable for all GL threads. Ideal for holding and storing gas-producing liquids, particularly hazardous chemicals. The gas-permeable membrane screw cap reduces the risk of over pressure and helps to stop a vacuum from forming. Membrane prevents infiltration by liquids and dirt particles.