You Searched For: Spreaders
54  results were found
SearchResultCount:"54"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Inoculating loops and needles are devices used for inoculation, serial dilution, sterile sampling, transfer, and microbiological sample spreading.
New Product
Supplier: VWR Collection
Description: PP, sterile (individually wrapped) or non sterile (bulk packed). Convenient and economical, these cell spreaders are designed to simplify bacterial cu...
Supplier: VWR Collection
Description: PS, sterile. These loops ensure consistent wetting and complete liquid transfer. Available rigid or flexible and have smooth edges to avoid gouging th...
Catalog Number: (612-9360)
Supplier: VWR Collection
Description: PS, sterile.
UOM: 1 * 1.000 items
Supplier: VWR Collection
Description: PS, sterile. Designed for spreading and dispersing liquids across the surface of agar culture plates. Used for performing bacterial (for example,...
Supplier: VWR Collection
Description: PS, sterile. Disposable plastic inoculating needles.
Supplier: VWR Collection
Description: These PS, sterile, disposable L-shaped spreaders enable even spreading of liquid samples across the surface of agar plates without gouging or cutting ...
Supplier: VWR Collection
Description: PS, sterile. The foot of the spreader has a completely smooth, rounded surface, free of rough edges and imperfections. It enables even spreading of li...
Supplier: VWR Collection
Description: These PS, sterile, disposable T-shaped spreaders are designed for spreading and dispersing liquids across the surface of agar culture plates. Their de...
Catalog Number: (612-2496)
Supplier: VWR Collection
Description: PS, sterile. Calibrated, disposable inoculating loops made from flexible or rigid plastic that cater for microbiologist's different applications and m...
UOM: 1 * 1.000 items
Supplier: COPAN
Description: These nickel-chrome metal wire loops are available non calibrated in a range of sizes with internal diameters (Ø) of 2, 3, 4 or 5 mm, or calibrated, 1...
Supplier: COPAN
Description: A range of nickel-chrome metal wire loops attached to lightweight insulated aluminium handles. Combo-loops are non calibrated and available in a range...
Supplier: RSG
Description: These NiCr 8020 loops are available in a range of sizes and lengths with internal diameter of 1,5, 2,5 or 4 mm. They are heat resistant and can be fla...
Supplier: 3M Food Safety
Description: This 3Mâ„¢ Petrifilmâ„¢ Spreader is an easy-to-use tool designed to quickly and evenly distribute inoculum within the 3Mâ„¢ Petrifilmâ„¢ Plate.
Supplier: COPAN
Description: Inoculating loop, 1 µl, Sterile
Supplier: BOCHEM
Description: Stainless steel 18/10. Length 100 mm.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 54
no targeter for Bottom