You Searched For: Cell Scrapers
12  results were found
SearchResultCount:"12"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Flat blade cell lifter for manual harvesting of cells, manufactured from high grade PE. Designed for removing cells from flasks or dishes.
Supplier: VWR Collection
Description: Specifically designed to make collecting cells easier and more effective. Available with a choice of blade positions: scraper for use in flasks, or li...
Supplier: Greiner Bio-One
Description: Designed to facilitate the gentle mechanical removal of adherent cells from growth surfaces. Each scraper has a 18 mm pivoted (pivot angle 60°) scrape...
Supplier: Corning
Description: PS handle, TPE blade. Designed to provide maximum accessibility to the growth surfaces of a variety of culture vessels.
Catalog Number: (231-2475)
Supplier: DELTALAB
Description: Wooden spatula with rounded edges. Non sterile.
UOM: 1 * 500 items
Supplier: Thermo Fisher Scientific
Description: Cell scrapers for cell harvesting are available in two lengths, with adjustable blade for optimal application flexibility. Non-pyrogenic.
Supplier: DELTALAB
Description: PP spatula with rounded edges. Sterile model is sterilised by ethylene oxide. Pack includes instructions for use.
Supplier: Corning
Description: Useful for the manual harvesting of cells, scrapers are designed for use in flasks and cell lifters for use in harvesting cells (especially stem cells...
Catalog Number: (22003.)
Supplier: Biotium
Description: PE, disposable, sterile. Mini cell scrapers for harvesting cells or cell lysates from microwell plates.
UOM: 1 * 200 items
Catalog Number: (WITG8.486.200)
Supplier: WITEG LABORTECHNIK GMBH
Description: Bent, plastic coated glass handle, mobile PTFE blade. Scraper for removal of cells from flasks and plates. Length 50 mm.
UOM: 1 * 1 items
Supplier: BELLCO GLASS INC.
Description: Stainless steel blade holder mounted on plastic covered stainless steel handle.
Catalog Number: (SARS83.3951)
Supplier: SARSTEDT
Description: Cell scraper with soft, flexible blade for gentle removal of monolayer cultures, minimising cell damage.
UOM: 1 * 100 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 12 of 12
no targeter for Bottom