You Searched For: Calipers
7  results were found
SearchResultCount:"7"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Made from stainless steel. Battery life is about 10000 hours.
Catalog Number: (819-0014)
Supplier: BOCHEM
Description: Caliper with five digit LCD.
UOM: 1 * 1 items
Catalog Number: (238-2100)
Supplier: BOCHEM
Description: Stainless steel Vernier caliper with black engraved scale.
UOM: 1 * 1 items
Catalog Number: (BELAH134170000)
Supplier: Bel-Art Products, a Part of SP
Description: Measurements taken with push button selection between metric and English scales and a self-zeroing feature for accuracy of ±0,1 mm.
UOM: 1 * 1 items
Catalog Number: (BELAH134150000)
Supplier: Bel-Art Products, a Part of SP
Description: Measure internal and external dimensions with high precision. Upper scale is 150 mm with 0,1 mm subdivisions, lower scale is 6" with 0,005" subdivisio...
UOM: 1 * 1 items
Catalog Number: (BELAH134160001)
Supplier: Bel-Art Products, a Part of SP
Description: This dial caliper can measure up to 150 mm on a white dial graduated in 0,1 mm increments. Vernier model measures 130 mm in 0,05 subdivisions, plus 5"...
UOM: 1 * 1 items
Catalog Number: (MOIN1452MI)
Supplier: MOINEAU
Description: Analogue caliper with clamping screw.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 7 of 7
no targeter for Bottom