Automatic burettes, pellet pattern, with intermediate Stopcock, SILBERBRAND
Supplier: Brand
SILBERBRAND
612-9917EA 269.7 EUR
612-9917
Automatic burettes, pellet pattern, with intermediate Stopcock, SILBERBRAND
Burettes
Borosilicate glass 5.4, amber, class B.
  • Calibrated to deliver (TD, Ex)
  • Short marks at major graduations, for 25 ml reduced distance between subdivision marks
  • Automatic zeroing and intermediate stopcock to recirculate residual liquid
25 ml: Error limits according to class B, DIN EN ISO 385.
50 ml: DIN EN ISO 385, class B.
*Total height includes the reservoir bottle.
Delivery information: Supplied with 2 litre (soda-lime glass) reservoir bottle.