You Searched For: Burettes
99  results were found
SearchResultCount:"99"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Borosilicate 3.3 glass, class AS.
Supplier: VWR Collection
Description: Borosilicate 3.3 glass, class AS.
Supplier: VWR Collection
Description: Borosilicate 3.3 glass, class AS.
Supplier: VWR Collection
Description: Borosilicate 3.3 glass, class B.
Supplier: VWR Collection
Description: Borosilicate 3.3 glass, class AS.
Supplier: VWR Collection
Description: Borosilicate 3.3 glass, class AS.
Catalog Number: (613-7002)
Supplier: VWR Collection
Description: State-of-the art motor-operated burette with three calibrated pre-set speeds and accuracy with precision conforming to ISO 8655 standards.
UOM: 1 * 1 items
Supplier: Brand
Description: Variable volume.
Low Manufacturing Impact
Sustainable Packaging
Supplier: Brand
Description: Borosilicate glass 3.3, class AS.
Supplier: Brand
Description: Borosilicate glass 3,3, PTFE spindle, class AS.
Supplier: Brand
Description: AR-GLAS®, class B.
Supplier: Brand
Description: Borosilicate glass 3.3, class AS.
Supplier: Thermo Fisher Scientific
Description: Crystal-clear acrylic, PMP tip and TFE stopcock and plug.
Sale
Supplier: Thermo Fisher Scientific
Description: Crystal-clear acrylic, PMP tip and TFE stopcock and plug, LDPE bottle with PP screw closure, 1 m PVC tubing.
Supplier: Brand
Description: AR-Glas®, class B.
Supplier: Brand
Description: Borosilicate glass 3.3, class AS, DE-M marking.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 99
no targeter for Bottom