VWR®, Cleaning Brushes for Glass Beakers
Supplier: VWR
VWR®
149-0131EA 14.3 EUR
149-0131 149-0132 149-0133
VWR®, Cleaning Brushes for Glass Beakers
Brushes
Natural black bristles with wooden handle. Inexpensive brushes for cleaning glass beakers and a wide range of other wide necked laboratory utensils.
  • Chemical resistant bristles
  • Long wooden handle
  • Hanging loop