Save on sample storage essentials
Automate your storage workflow and save with Thermo Scientificâ„¢ sample storage starter packs. Choose a pack to suit your lab and help boost productivity.
Shop now
Save now
Learn more
You Searched For: Boxes
361  results were found
SearchResultCount:"361"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Cardboard.
Supplier: VWR Collection
Description: Cardboard, coated.
Supplier: VWR Collection
Description: PS, clear.
Supplier: VWR Collection
Description: Insulated waterproof carrier.
Supplier: VWR Collection
Description: A broad variety of PP storage boxes to securely hold a wide range of vial sizes and bottles. Perfect for space-saving storage in the laboratory or in ...
Supplier: VWR Collection
Description: Corrugated cardboard box.
Supplier: VWR Collection
Description: Cryo boxes are suitable to store up to –196 °C.
Sale
Supplier: VWR Collection
Description: These sturdy, transparent plastic containers are made of translucent material. The translucent material allows users to check whether a container is e...
Supplier: VWR Collection
Description: Cardboard, coated.
Supplier: VWR Collection
Description: Stainless steel, with square base, autoclavable.
Supplier: VWR Collection
Description: Cardboard, coated.
Supplier: VWR Collection
Description: Made of extra strong polycarbonate.
Supplier: VWR Collection
Description: PP.
Supplier: VWR Collection
Description: Cardboard, white.
Catalog Number: (631-1050)
Supplier: VWR Collection
Description: Storage organiser with 2×4 metal drawers for embedding cassettes.
UOM: 1 * 1 items
Supplier: VWR Collection
Description: This range of plastic boxes offers a modern and practical solution for the storage, organization, transportation, and presentation of small parts. Wit...
New Product Sale
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 361
no targeter for Bottom