VWR®, Bottles, Wide Neck, Square, with Screw Cap
Supplier: VWR
215-7396EA 9.1 EUR
215-7396 215-7390 215-7392 215-7394
VWR®, Bottles, Wide Neck, Square, with Screw Cap
Bottles Plastic Bottles
HDPE with black, HDPE, tamper-evident cap.
  • Good chemical resistance
  • Leakproof
  • Space saving
Perfect for sample collection, storage and transport. For liquids, pastes and granular material. The wide mouth allows easy filling and cleaning.