You Searched For: Bottles
1 126  results were found
SearchResultCount:"1126"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: VWR® sample bottles are ideal for storage of media, laboratory reagents, and biological buffers in temperatures ranging from −70 to 60 °C. Eliminate w...
New Product
Supplier: VWR Collection
Description: LDPE, amber.
Supplier: VWR Collection
Description: Square PETG bottles are ideal for storage of media, laboratory reagents and biological buffers up to −70 °C. Eliminate washing, depyrogenation and aut...
New Product
Supplier: Avantor Fluid Handling
Description: Safely transfer, sample, and store biopharmaceutical products and reagents.
Supplier: Avantor Fluid Handling
Description: Safely transfer, sample, and store biopharmaceutical products and reagents.
Supplier: VWR Collection
Description: Bottles are made with HDPE resin.
Supplier: VWR Collection
Description: HDPE or PP translucent, with purple screw caps.
Supplier: VWR Collection
Description: HDPE water sampling collection bottle.
Supplier: VWR Collection
Description: PET, with or without PP screw caps.
Supplier: VWR Collection
Description: LDPE/HDPE or PP, translucent with purple screw caps. Use for storing and dispensing solutions and media. Round carboys feature an integrated shoulder ...
Supplier: VWR Collection
Description: PTFE, white.
Supplier: VWR Collection
Description: Wide neck square bottles available in HDPE or PP material.
Catalog Number: (216-1917)
Supplier: VWR Collection
Description: Disposable jerrican, made of HDPE, with integrated handle for easy carrying and pouring.
UOM: 1 * 1 items
Supplier: VWR Collection
Description: PET, virgin, food grade with PP caps.
Supplier: VWR Collection
Description: Integrated safety vent cap eliminates dripping of vapours.
Sale
Supplier: VWR Collection
Description: LDPE, translucent, with coloured, PP spray cap.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 1 126
no targeter for Bottom